Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP64
Entrez Gene ID
485932
Gene Name
ZFP64 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.24: 38201824 - 38289395 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639867.4 XP_003639915.1 3 - 4 627 144
XM_005635186.3 XP_005635243.1 3 - 4 444 144
XM_014107191.2 XP_013962666.1 4 - 5 710 144
XM_862201.4 XP_867294.2 3 - 4 444 144

View Full Product Details