Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BBC3
Entrez Gene ID
612153
Gene Name
BCL2 binding component 3
Gene Aliases
-
Chromosome Location
Chr.1: 108742793 - 108750832 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616403.3 XP_005616460.1 4 - 4 2247 68
XM_022424594.1 XP_022280302.1 5 - 5 657 68
XM_849883.5 XP_854976.1 4 - 4 768 68

View Full Product Details