Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
SCD5
Entrez Gene ID
487831
Gene Name
stearoyl-CoA desaturase 5
Gene Aliases
-
Chromosome Location
Chr.32: 6594607 - 6745379 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544953.6 XP_544953.2 4 - 4 956 121

View Full Product Details