Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPTY2D1
Entrez Gene ID
476884
Gene Name
SPT2 chromatin protein domain containing 1
Gene Aliases
-
Chromosome Location
Chr.21: 41021317 - 41042955 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022407918.1 XP_022263626.1 4 - 4 932 186
XM_022407919.1 XP_022263627.1 3 - 3 674 186
XM_534086.5 XP_534086.3 3 - 3 769 186

View Full Product Details