Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXCR4
Entrez Gene ID
483900
Gene Name
C-X-C motif chemokine receptor 4
Gene Aliases
-
Chromosome Location
Chr.19: 38874650 - 38877740 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048026.1 NP_001041491.1 2 - 2 948 137
GenBank mRNA DQ182699.1 - 2 - 2 948 137

View Full Product Details