Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT76
Entrez Gene ID
486518
Gene Name
keratin 76
Gene Aliases
KRT2P
Chromosome Location
Chr.27: 2337929 - 2346716 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001346020.1 NP_001332949.1 4 - 5 1024 72

View Full Product Details