Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT9
Entrez Gene ID
490980
Gene Name
keratin 9
Gene Aliases
CK-9, K9
Chromosome Location
Chr.9: 21204213 - 21210793 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022423953.1 XP_022279661.1 - 993 70

View Full Product Details