Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPTL1
Entrez Gene ID
612843
Gene Name
angiopoietin like 1
Gene Aliases
-
Chromosome Location
Chr.7: 20919192 - 20945140 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109813.1 NP_001103283.1 2 - 3 501 123

View Full Product Details