The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109958.1 | NP_001103428.1 | 1 - 2 | 200 | 122 |
GenBank mRNA | AM748733.1 | - | 1 - 2 | 200 | 122 |