Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GYPA
Entrez Gene ID
100126181
Gene Name
glycophorin A (MNS blood group)
Gene Aliases
-
Chromosome Location
Chr.15: 43018705 - 43049726 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109960.1 NP_001103430.1 2 - 3 166 95
GenBank mRNA AB299408.1 - 2 - 3 166 95

View Full Product Details