Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL29L
Entrez Gene ID
611377
Gene Name
interleukin 29-like
Gene Aliases
-
Chromosome Location
Chr.24: 36440445 - 36441017 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114853.1 NP_001108325.1 1 - 1 448 143
GenBank mRNA AB241609.1 - 1 - 1 448 143
KC754970.1 - 1 - 1 448 143

View Full Product Details