Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFSF8
Entrez Gene ID
612608
Gene Name
TNF superfamily member 8
Gene Aliases
TNLG3A
Chromosome Location
Chr.11: 68979498 - 69006898 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850342.4 XP_855435.1 3 - 4 629 102

View Full Product Details