The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128064.1 | NP_001121536.1 | - | 947 | 113 |
GenBank mRNA | AB436535.1 | - | - | 947 | 113 |