Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INHBC
Entrez Gene ID
607422
Gene Name
inhibin subunit beta C
Gene Aliases
-
Chromosome Location
Chr.10: 1546654 - 1558631 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844076.5 XP_849169.2 - 737 108

View Full Product Details