Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIIAD1
Entrez Gene ID
100294631
Gene Name
regulatory subunit of type II PKA R-subunit domain containing 1
Gene Aliases
-
Chromosome Location
Chr.17: 60753380 - 60759713 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159762.2 NP_001153234.1 3 - 4 346 80
GenBank mRNA CX993709.1 - 1 - 2 125 80
DN395231.1 - 3 - 4 318 80

View Full Product Details