Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELENOI
Entrez Gene ID
100302747
Gene Name
selenoprotein I
Gene Aliases
EPT1, SELI
Chromosome Location
Chr.17: 20431176 - 20469694 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164486.1 NP_001157958.1 6 - 7 802 106

View Full Product Details