Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC25
Entrez Gene ID
481665
Gene Name
DnaJ heat shock protein family (Hsp40) member C25
Gene Aliases
ERJ7, GNG10
Chromosome Location
Chr.11: 66234583 - 66261153 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205003.1 NP_001191932.1 2 - 3 543 75

View Full Product Details