Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM34
Entrez Gene ID
100568302
Gene Name
tripartite motif containing 34
Gene Aliases
-
Chromosome Location
Chr.21: 28635177 - 28656759 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205191.1 NP_001192120.1 1 - 2 97 104
XM_022407408.1 XP_022263116.1 1 - 2 316 104
XM_022407409.1 XP_022263117.1 1 - 2 314 104
GenBank mRNA DN267373.1 - 1 - 2 92 104

View Full Product Details