Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEF2B
Entrez Gene ID
609739
Gene Name
myocyte enhancer factor 2B
Gene Aliases
-
Chromosome Location
Chr.20: 44000253 - 44017371 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205203.1 NP_001192132.1 6 - 7 773 67
XM_022406260.1 XP_022261968.1 7 - 8 1679 67
XM_022406261.1 XP_022261969.1 7 - 8 942 67

View Full Product Details