Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL14
Entrez Gene ID
477031
Gene Name
ribosomal protein L14
Gene Aliases
-
Chromosome Location
Chr.23: 9907479 - 9911114 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252411.1 NP_001239340.1 5 - 6 406 89
GenBank mRNA GR886597.1 - 5 - 6 406 89

View Full Product Details