Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAVCR2
Entrez Gene ID
479318
Gene Name
hepatitis A virus cellular receptor 2
Gene Aliases
-
Chromosome Location
Chr.4: 53018990 - 53032035 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254715.1 NP_001241644.1 4 - 5 517 69

Gene Symbol
HAVCR1
Entrez Gene ID
612069
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
-
Chromosome Location
Chr.4: 53012223 - 53066611 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022417343.1 XP_022273051.1 6 - 7 960 69
XM_022417344.1 XP_022273052.1 6 - 7 840 69
XM_022417347.1 XP_022273055.1 5 - 6 900 69

View Full Product Details