Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRDX3
Entrez Gene ID
477839
Gene Name
peroxiredoxin 3
Gene Aliases
AOP1
Chromosome Location
Chr.28: 29395593 - 29403745 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256485.1 NP_001243414.1 2 - 3 196 60

View Full Product Details