Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR27B
- Entrez Gene ID
-
100886115
- Gene Name
- microRNA mir-27b
- Gene Aliases
-
cfa-mir-27b
- Chromosome Location
-
Chr.1: 71678117 - 71678179 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_049240.1 |
|
1 - 1 |
32 |
63 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
spalt like transcription factor 3 |
SALL3 |
|
-701.8kb
|
Sense |
Chr. - 1413991 - 1421516 on Build CanFam3.1 |
Downstream |
uncharacterized LOC111092046 |
LOC111092046 |
|
179.9kb
|
Anti-sense |
Chr. - 2303295 - 2305318 on Build CanFam3.1 |
View Full Product Details