Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR18B
- Entrez Gene ID
-
100885977
- Gene Name
- microRNA mir-18b
- Gene Aliases
-
cfa-mir-18b
- Chromosome Location
-
Chr.X: 104896339 - 104896415 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_049417.1 |
|
1 - 1 |
41 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
glypican 3 |
GPC3 |
|
-49.6kb
|
Sense |
Chr. - 104313426 - 104751215 on Build CanFam3.1 |
Downstream |
nuclease EXOG, mitochondrial-like |
LOC100686111 |
|
56.2kb
|
Anti-sense |
Chr. - 104857047 - 104858004 on Build CanFam3.1 |
View Full Product Details