Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR33B
- Entrez Gene ID
-
100885995
- Gene Name
- microRNA mir-33b
- Gene Aliases
-
cfa-mir-33b
- Chromosome Location
-
Chr.5: 41685165 - 41685258 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_049451.1 |
|
1 - 1 |
52 |
52 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
target of myb1 like 2 membrane trafficking protein |
TOM1L2 |
|
-30.5kb
|
Sense |
Chr. - 41535069 - 41654609 on Build CanFam3.1 |
View Full Product Details