Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- H19
- Entrez Gene ID
-
100271858
- Gene Name
- H19, imprinted maternally expressed transcript (non-protein coding)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 46204871 - 46207573 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027327.2 |
|
1 - 2 |
1419 |
55 |
GenBank mRNA |
FJ159122.1 |
- |
1 - 2 |
653 |
55 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
semaphorin 3C |
SEMA3C |
|
-125.9kb
|
Sense |
Chr. - 20457683 - 20748008 on Build CanFam3.1 |
Downstream |
hepatocyte growth factor |
HGF |
|
454.2kb
|
Sense |
Chr. - 21329688 - 21410257 on Build CanFam3.1 |
View Full Product Details