Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cacng5
Entrez Gene ID
100766301
Gene Name
calcium channel, voltage-dependent, gamma subunit 5
Gene Aliases
I79_008282
Chromosome Location
Chr.Un|NW_003613864: 130679 - 137626 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501852.1 XP_003501900.1 4 - 5 573 67

View Full Product Details