Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
March3
Entrez Gene ID
100752992
Gene Name
membrane-associated ring finger (C3HC4) 3, E3 ubiquitin protein ligase
Gene Aliases
I79_003939
Chromosome Location
Chr.Un|NW_003613682: 2112526 - 2146965 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498174.2 XP_003498222.2 1 - 2 257 58

View Full Product Details