Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spin1
Entrez Gene ID
100763151
Gene Name
spindlin 1
Gene Aliases
Spindlin-1
Chromosome Location
Chr.Un|NW_003615924: 35100 - 71647 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513430.2 XP_003513478.1 2 - 3 303 89

View Full Product Details