Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Layn
Entrez Gene ID
100689416
Gene Name
layilin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613694: 832380 - 850274 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246681.1 NP_001233610.1 3 - 4 655 87
XM_007629041.1 XP_007627231.1 4 - 5 821 87
GenBank mRNA AF093673.1 - 3 - 4 679 87

View Full Product Details