Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Neu1
Entrez Gene ID
100689373
Gene Name
sialidase 1 (lysosomal sialidase)
Gene Aliases
I79_016442
Chromosome Location
Chr.Un|NW_003614548: 321895 - 324224 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246800.1 NP_001233729.1 2 - 3 606 99
GenBank mRNA FJ476073.1 - 2 - 3 606 99
JI895720.1 - 2 - 3 367 99
JP057040.1 - 2 - 3 367 99

View Full Product Details