Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhfr
Entrez Gene ID
100689028
Gene Name
dihydrofolate reductase
Gene Aliases
I79_015631
Chromosome Location
Chr.Un|NW_003614442: 464809 - 488594 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244016.1 NP_001230945.1 3 - 4 248 109
GenBank mRNA JI884329.1 - 3 - 4 341 109
JP060064.1 - 3 - 4 341 109
K01164.1 - 3 - 4 248 109

View Full Product Details