Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps17
Entrez Gene ID
100689053
Gene Name
ribosomal protein S17
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615555: 133078 - 135550 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244040.1 NP_001230969.1 1 - 2 172 105
GenBank mRNA JI881027.1 - 2 - 3 193 105
JP060743.1 - 2 - 3 193 105
M13933.1 - 1 - 2 172 105

View Full Product Details