Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cga
Entrez Gene ID
100689452
Gene Name
glycoprotein hormones, alpha polypeptide
Gene Aliases
I79_010669
Chromosome Location
Chr.Un|NW_003614000: 865507 - 877147 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246711.1 NP_001233640.1 2 - 3 208 116
GenBank mRNA AB248597.1 - 2 - 3 208 116

View Full Product Details