Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dph2
Entrez Gene ID
100689365
Gene Name
DPH2 homolog (S. cerevisiae)
Gene Aliases
I79_009299
Chromosome Location
Chr.Un|NW_003613906: 171350 - 173562 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246793.1 NP_001233722.1 4 - 5 1345 84
GenBank mRNA DQ981502.1 - 4 - 5 1345 84

View Full Product Details