Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdc1
Entrez Gene ID
100689057
Gene Name
syndecan 1
Gene Aliases
I79_008577, SYND1
Chromosome Location
Chr.Un|NW_003613876: 572441 - 574800 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244042.1 NP_001230971.1 2 - 3 625 73
GenBank mRNA JI895124.1 - 2 - 3 600 73
JP063507.1 - 2 - 3 600 73
L38991.1 - 2 - 3 625 73

View Full Product Details