Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Icam1
Entrez Gene ID
100689428
Gene Name
intercellular adhesion molecule 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613752: 1193507 - 1205407 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246690.1 NP_001233619.1 2 - 3 486 115
GenBank mRNA AF308456.1 - 2 - 3 486 115
JI866855.1 - 2 - 3 407 115
JP055129.1 - 2 - 3 407 115

View Full Product Details