Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbx1
Entrez Gene ID
100689359
Gene Name
chromobox homolog 1
Gene Aliases
I79_005678
Chromosome Location
Chr.Un|NW_003613745: 619094 - 637494 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246787.1 NP_001233716.1 - 353 76
GenBank mRNA AY548739.1 - - 353 76
JI876171.1 - 2 - 3 369 76
JP059074.1 - 2 - 3 369 76

View Full Product Details