Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dio3
Entrez Gene ID
100753739
Gene Name
deiodinase, iodothyronine, type III
Gene Aliases
I79_015795
Chromosome Location
Chr.Un|NW_003614463: 580642 - 582214 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256784.1 NP_001243713.1 1 - 1 1020 71

View Full Product Details