Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35b4
Entrez Gene ID
100689030
Gene Name
solute carrier family 35 (UDP-xylose/UDP-N-acetylglucosamine transporter), member B4
Gene Aliases
Slc25b4
Chromosome Location
Chr.Un|NW_003613945: 1356197 - 1375109 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244018.2 NP_001230947.1 1 - 2 265 79
GenBank mRNA FN825778.1 - 1 - 2 269 79
JI894393.1 - 1 - 2 177 79
JP044451.1 - 1 - 2 177 79

View Full Product Details