Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc35c1
Entrez Gene ID
100689382
Gene Name
solute carrier family 35 (GDP-fucose transporter), member C1
Gene Aliases
I79_002855
Chromosome Location
Chr.Un|NW_003613646: 1209624 - 1216434 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246808.1 NP_001233737.1 1 - 2 536 73
XM_007609797.1 XP_007607987.1 2 - 3 656 73
GenBank mRNA AB222037.1 - 1 - 2 536 73

View Full Product Details