Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efemp2
Entrez Gene ID
100736553
Gene Name
EGF containing fibulin-like extracellular matrix protein 2
Gene Aliases
FBLN4, FIBL-4, Fibulin-4, H411, I79_017609
Chromosome Location
Chr.Un|NW_003614712: 528661 - 535794 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244849.1 NP_001231778.1 9 - 10 1467 88
GenBank mRNA AF046870.1 - 9 - 10 1467 88

View Full Product Details