Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bak1
Entrez Gene ID
100689368
Gene Name
BCL2-antagonist/killer 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614972: 220625 - 229089 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246795.1 NP_001233724.1 2 - 3 203 98
XM_007651403.1 XP_007649593.1 3 - 4 1039 98
XM_007651404.1 XP_007649594.1 3 - 4 1022 98
GenBank mRNA EF104644.1 - 2 - 3 203 98
JI867360.1 - 3 - 4 481 98
JP056774.1 - 3 - 4 480 98

View Full Product Details