Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hsp90aa1
Entrez Gene ID
100689397
Gene Name
heat shock protein 90kDa alpha (cytosolic), class A member 1
Gene Aliases
HSPCA, Hsp90a, I79_004665
Chromosome Location
Chr.Un|NW_003613702: 274695 - 279361 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246821.1 NP_001233750.1 1 - 2 181 98
GenBank mRNA JI879574.1 - 1 - 2 213 98
JP066070.1 - 1 - 1 213 98
L33676.1 - 1 - 2 181 98

View Full Product Details