Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phospho2
Entrez Gene ID
100764721
Gene Name
phosphatase, orphan 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613606: 988776 - 991778 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495941.2 XP_003495989.1 1 - 1 617 98
XM_007645399.1 XP_007643589.1 2 - 2 712 98

View Full Product Details