Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxl14
Entrez Gene ID
100755365
Gene Name
F-box and leucine-rich repeat protein 14
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613787: 908908 - 912325 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500330.2 XP_003500378.1 1 - 2 203 66

View Full Product Details