Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wasf2
Entrez Gene ID
100759260
Gene Name
WAS protein family, member 2
Gene Aliases
I79_019962
Chromosome Location
Chr.Un|NW_003615112: 241217 - 286580 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651967.1 XP_007650157.1 6 - 7 729 74

View Full Product Details