Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc85c
Entrez Gene ID
100769004
Gene Name
coiled-coil domain containing 85C
Gene Aliases
I79_010593
Chromosome Location
Chr.Un|NW_003613993: 484703 - 551751 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503715.2 XP_003503763.1 3 - 4 248 62

View Full Product Details