Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ncr3
Entrez Gene ID
100772283
Gene Name
natural cytotoxicity triggering receptor 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614548: 566557 - 570244 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508508.1 XP_003508556.1 1 - 2 29 66

View Full Product Details