Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Elfn1
Entrez Gene ID
100770773
Gene Name
extracellular leucine-rich repeat and fibronectin type III domain containing 1
Gene Aliases
I79_001696
Chromosome Location
Chr.Un|NW_003613618: 3799549 - 3858002 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496322.1 XP_003496370.1 3 - 4 245 65

View Full Product Details